missing translation for 'onlineSavingsMsg'
Learn More

Water, MilliporeSigma™

Catalog No. MWX00046
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MWX00046 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MWX00046 Supplier Technidata Supplier No. WX00046

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

Molecular Formula H2O
Synonym Aqua-Destillata

Specifications

Linear Formula H2O
Formula Weight 18.02
Packaging Glass Bottle
Quantity 1 L
Chemical Name or Material LC-MS Water