missing translation for 'onlineSavingsMsg'
Learn More

Water ACS reagent, For ultratrace analysis, MilliporeSigma™ Supelco™

Catalog No. 111015496
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015496 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015496 Supplier Merck Emd Millipore Supplier No. 142111LF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00011332

Specifications

CAS 7732-18-5
Density 1.000 g/mL (at 3.98°C)
Boiling Point 100°C
Linear Formula H2O
Quantity 1 L
Formula Weight 18.02
Grade ACS Reagent
RTECSNumber ZC0110000