missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma Water

Catalog No. 01257782
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01257782 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01257782 Supplier MilliporeSigma Supplier No. 1047351000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Water tested for EPA 533 and EPA 537.1 PFAS methods LiChrosolv™.

  • PFAS tested solvents are used for UHPLC-MS and LC-MS instrumental analysis.

Specifications

Chemical Name or Material Water
CAS 7732-18-5
Molecular Weight (g/mol) 18.02 g/mol
Grade UHPLC-MS
Packaging Borosilicate Glass Bottle
Quantity 1 L