missing translation for 'onlineSavingsMsg'
Learn More

Water with 0.1% ammonium acetate, Solstice

Catalog No. 60048249
Change view
Click to view available options
Quantity:
4 x 4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60048249 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60048249 Supplier Solstice Supplier No. 346744X4L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CHROMASOLV™ LC-MS, LabReady™ Solvent Blend

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities

Specifications

Packaging Glass Bottle
Quantity 4 x 4 L