Learn More
Water with 0.1% acetic acid, Honeywell Riedel-de Haën™
CHROMASOLV™ LC-MS, LabReady™ Solvent Blend
$1785.78 - $1785.78
Specifications
Packaging Glass Bottle
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60048248
View Documents
Honeywell Chemicals
346754X4L
4 x 4 L Glass Bottle
Case of 4 Each for $1,785.78
Only null left
Description
Description
  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities
Specifications
Specifications
Glass Bottle
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'