Learn More
Electron Microscopy Sciences Wash N'Dry™ Wash N'Dry Cover Slip Rack
Supplier:  Electron Microscopy Sciences 7036616
Encompass
Cover slip rack that holds 10 cover slips.Fast and easy cleaning, surface-treating and drying. Holds all slipcover sizes including 18, 22 and 25 mm. One piece polypropylene rack does not require assembly and is chemically and thermally resistant.Autoclavable. Innovative design includes a notch to secure coverslips with a rubber band or loop of monofilament. Fits inside a 100 ml beaker with space underneath for a small stir bar when submerged.
Catalog No. 5034535
Only null left