missing translation for 'onlineSavingsMsg'
Learn More

ShockWatch™ WarmMark™ Time-Temperature Indicator

Catalog No. 22111030
Encompass
Change view
Click to view available options
Temperature Range (English):
36°F
46°F
Measurement Time:
48 hr.
8 hr.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (English) Measurement Time
22111030 46°F 8 hr.
22111029 46°F 48 hr.
22111031 36°F -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Contains no known hazardous or carcinogenic components
Catalog No. 22111030 Supplier ShockWatch™ Supplier No. WM 8/468

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Contains no known hazardous or carcinogenic components

ShockWatch WarmMark Temperature Indicator is a single-use dacending temperature indicator, which alerts users of exposure to unacceptable temperature conditions.

  • Field armable with pull tab
  • Black stripes show product armed
  • Brief, moderate, prolonged exposure times

Order Info

Store in dry environment below response temperature for optimal shelf life

Certifications

ROHS2, REACH

Warranty

2 years from date of sale

Specifications

Certifications/Compliance ROHS2, REACH
Disposable Single-use
For Use With (Application) Shipping and storage
Measurement Time 8 hr.
Thermometer Fill White to Red
Product Type Disposable Temperature Indicator
Max. Temperature (English) 46°F
Max. Temperature (Metric) 8°C
Min. Temperature (English) 46°F
Min. Temperature (Metric) 8°C
Temperature Range (English) 46°F
Temperature Range (Metric) 8°C
Accuracy ±1°C
Show More Show Less