Learn More
Conair™ Waring™ Variable Speed Control for Blender
Catalog No. 1450538
Change view
Click to view available options
For Use With (Equipment):
Waring™ blender
Catalog No. For Use With (Equipment)
1450538 Waring™ blender
1 options
Catalog No. 1450538 Supplier Conair™ Supplier No. LA1
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Provides infinite speed control (0 to unit maximum) for blenders up to 8A maximum. With ON/OFF switch, speed adjustment control, 3-wire grounded cord. 120V 50/60Hz, 8A.
Specifications
Product Type Speed Controller
For Use With (Application) Ideal for provides infinite speed control
For Use With (Equipment) Waring™ blender