missing translation for 'onlineSavingsMsg'
Learn More

Conair™ Waring™ Laboratory Blenders: Two Speeds with Timer

Catalog No. p-4906756
Change view
Click to view available options
Dimensions (L x W x H):
9.75 x 8 x 15 in.
9.75 x 8 x 16 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
145097P 9.75 x 8 x 15 in.
145097M 9.75 x 8 x 16 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 145097P Supplier Conair™ Supplier No. 7010S
Only null left

Includes

Base, container, 3-wire grounded cord and plug

Dual-speed action for demanding mixing and blending applications

  • Cloverleaf design
  • Choice of glass and/or stainless-steel containers
  • Sturdy, diecast metal bases and stainless-steel blades
  • All durably constructed for years of dependable service
  • Epoxy-coated bases with chrome top pieces on all models, except for the aluminum and steel base (Hazardous-Location Model)
  • Heat-resistant borosilicate glass (integral handle) or stainless-steel (attached handles)
  • Two-piece vinyl and styrene lids

Order Info

Electrical requirements for all models: 115/120V 50/60Hz

Certifications

UL listed (Select models only; see ordering chart)

Specifications

Amperage 3.6 A
Speed 18,000, 22,000 rpm
Voltage 115/120 V
Electrical Requirements 115/120 V, 50/60 Hz
Hertz 50/60 Hz
Horsepower 0.4 hp
Capacity (Metric) 1000 mL
Sample Volume (English) 34 oz.
Dimensions (L x W x H) 9.75 x 8 x 15 in.
Height (English) 15 in.
Height (Metric) 38 cm
Length (English) 9.75 in.
Length (Metric) 24.76 cm
Width (English) 8 in.
Width (Metric) 20.32 cm
Base Material Epoxy-coated Metal
Container Material Stainless Steel
Includes Base, container, 3-wire grounded cord and plug, timer
Packaging Quantity 1 Ea.
Show More Show Less