Learn More
Eisco™ Wankel Rotary Engine Model
Catalog No. S13481
Change view
Click to view available options
8 x 7 x 3.5 in.
Catalog No.
S13481 8 x 7 x 3.5 in.
1 options
Catalog No. S13481 Supplier Eisco™ Supplier No. PH0496
Shows the internal structure and operating principles of a Wankel engine.
  • Cut away to show internal constructional details
  • Rotary piston engine avoids reciprocating parts
  • Power piston is arch-like triangular rotor which generates epitrechoid upon rotation
  • Hand-powered motion supplied through crank demonstrates moving parts of engine as handcrank is rotated
  • Simulated spark plug uses small lamp to indicate firing point
  • Constructed from aluminum and plastic
Specifications
Product Type Model, Engine, Wankel Rotary
Dimensions (L x W x H) 8 x 7 x 3.5 in.