Learn More
Contec™ Wall WipR™ Cleaning System and Accessories
Offers wide surface coverage and low operator fatigue
$192.98
Specifications
Sterility Non-sterile
Autoclavable Autoclavable
For Use With (Equipment) Contec™ Cleaning System
For Use With (Application) Cleaning system is designed to clean walls and ceilings in controlled environments
View More Specs
Products 1
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
19038571
View Documents Promotion Details
Contec™
WWC18
Mop Cover
Pack of 50 Each for $192.98
Case of 8 Pack for $1,543.84
Only null left
Description
Description
  • Polyester fabric combined with a lightweight aluminum frame and handle offers wide surface coverage and low operator fatigue
  • The specially-designed fabric covers are low in extractable residues and even lower in particle generation
  • Long, lightweight handle allows easy cleaning of ceilings and walls
  • Swivel “T” handle, 18 in. (45.7cm)
  • Polyester/foam wiping cover, 18 in. (45.7cm)
  • Aluminum extension handle, 8 ft. or 14 in.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.
Specifications
Specifications
Non-sterile
Autoclavable
Contec™ Cleaning System
Cleaning system is designed to clean walls and ceilings in controlled environments