Learn More
Sutter Instruments Thick / Standard Walled Borosilicate Glass, 250/pk
Catalog No. NC0259102
Encompass
Outer diameter 1.50mm, Inner Diameter 0..86mm, Overall length 10cm, 225/pk
Catalog No. NC0259102 Supplier Sutter Instruments Supplier No. BF1508610
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Outer diameter 1.50mm, Inner Diameter 0..86mm, Overall length 10cm, 225/pk