Learn More
Fujifilm Irvine WAKO EXTRACTION KIT DFS Item
Supplier:  Fujifilm Irvine Scientific 29550201
*Orders for this supplier are non-cancellable/non-returnable once they are processed.* DNA Extractor Kit 50TESTS Grade: for Genetic Research Storage condition: KEEP AT 2-10 DEGREES C Detects and quantitates contaminant DNA in serum and residual DNA in biopharmaceuticals
Catalog No. NC9419399
Only null left
Explore available promotions