missing translation for 'onlineSavingsMsg'
Learn More

Mopec™ Waffle Specimen Chucks for SHANDON Cryostats

Catalog No. 22445847
Change view
Click to view available options
Color:
Blue
Green
Orange
Pink
Purple
Red
White
Yellow
Diameter (Metric):
30 mm
40 mm
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Diameter (Metric)
22445847 Blue 30 mm
22445570 Green 30 mm
22445585 Orange 30 mm
22445862 Pink 30 mm
22445598 Purple 30 mm
22445771 Red 30 mm
22445826 White 30 mm
22445773 Yellow 30 mm
22445772 Blue 40 mm
22445724 Green 40 mm
22445624 Orange 40 mm
22445608 Pink 40 mm
22445673 Purple 40 mm
22445337 Red 40 mm
22445672 White 40 mm
22445365 Yellow 40 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 22445847 Supplier Mopec™ Supplier No. SP143WBL

Waffle Specimen Chucks are available in multiple sizes and colors.

  • Round, aluminum specimen chucks

Specifications

Product Type Waffle Chuck
For Use With (Equipment) SHANDON Cryostats
Diameter (Metric) 30 mm
Color Blue