missing translation for 'onlineSavingsMsg'
Learn More

Mopec™ Waffle Specimen Chucks for Leica, TBS, and Tanner Cryostats

Catalog No. 22445766
Change view
Click to view available options
Color:
Blue
Green
Orange
Pink
Purple
Red
White
Yellow
Diameter (Metric):
30 mm
40 mm
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Diameter (Metric)
22445766 Blue 30 mm
22445739 Green 30 mm
22445586 Orange 30 mm
22445449 Pink 30 mm
22445450 Purple 30 mm
22445262 Red 30 mm
22445870 White 30 mm
22445582 Yellow 30 mm
22445648 Blue 40 mm
22445759 Green 40 mm
22445850 Orange 40 mm
22445747 Pink 40 mm
22445851 Purple 40 mm
22445645 Red 40 mm
22445649 White 40 mm
22445544 Yellow 40 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 22445766 Supplier Mopec™ Supplier No. SP103WBL

Waffle Specimen Chucks are available in multiple sizes and colors.

  • Round, aluminum specimen chucks
  • Suitable for Leica (except CM1950 model), TBS, and Tanner Cryostats

Specifications

For Use With (Equipment) Leica, TBS, and Tanner Cryostats
Diameter (Metric) 30 mm
Color Blue