Learn More
Ted Pella Inc Wafer Cleaving / Glass Breaking Pliers, 6"", including jaw inserts
Numéro de catalogue. NC0169386
Encompass
The wafer cleaving / Glass Breaking pliers are used to make clean breaks of silicon wafers, glass, quartz or thin ceramic tiles. They are also called glass pliers or running pliers. The pliers come with two sets of jaw inserts: one soft set to break out over a the scored line. A white line on the nose of the pliers helps with alignment of the breaking line, one flat metal set nipping along a parallel score line.
Numéro de catalogue. NC0169386 Fournisseur Ted Pella Inc Code fournisseur TPI7295
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
The wafer cleaving / Glass Breaking pliers are used to make clean breaks of silicon wafers, glass, quartz or thin ceramic tiles. They are also called glass pliers or running pliers. The pliers come with two sets of jaw inserts: one soft set to break out over a the scored line. A white line on the nose of the pliers helps with alignment of the breaking line, one flat metal set nipping along a parallel score line.