Learn More
W.S. TYLER™ RO-TAP™ II Sieve Shaker Maintenance Kits
Catalog No. 0490914
Change view
Click to view available options
For Use With (Equipment):
RX-29/30 Ro-Tap Sieve Shaker
RX-29/30/94 Ro-Tap Sieve Shaker
RX-94 Ro-Tap sieve shakers
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
0490914 RX-29/30 Ro-Tap Sieve Shaker
0490913 RX-29/30/94 Ro-Tap Sieve Shaker
0490915 RX-94 Ro-Tap sieve shakers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 0490914 Supplier W.S. TYLER™ Supplier No. WST10122
Only null left
For replacement of parts due to normal wear and tear.
Specifications
Product Type Sieve Shaker Maintenance Kit
For Use With (Equipment) RX-29/30 Ro-Tap Sieve Shaker
Includes Timing belt, cam gear, timing belt pulley