missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VX-702 98.0+%, TCI America™
$281.08 - $841.30
Chemical Identifiers
| CAS | 745833-23-2 |
|---|---|
| Molecular Formula | C19H12F4N4O2 |
| Molecular Weight (g/mol) | 404.33 |
| MDL Number | MFCD11616590 |
| InChI Key | FYSRKRZDBHOFAY-UHFFFAOYSA-N |
| Synonym | 6-[(Aminocarbonyl)(2,6-difluorophenyl)amino]-2-(2,4-difluorophenyl)-3-pyridinecarboxamide, 2-(2,4-Difluorophenyl)-6-(1-[2,6-difluorophenyl)ureido]nicotinamide |
| PubChem CID | 10341154 |
| IUPAC Name | 2-(2,4-difluorophenyl)-6-[1-(2,6-difluorophenyl)carbamoylamino]pyridine-3-carboxamide |
| SMILES | NC(=O)N(C1=NC(=C(C=C1)C(N)=O)C1=C(F)C=C(F)C=C1)C1=C(F)C=CC=C1F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
V014725MG
|
TCI America
V014725MG |
25 mg |
Each for $281.08
|
|
|||||
|
V0147100MG
|
TCI America
V0147100MG |
100 mg |
Each for $841.30
|
|
|||||
Chemical Identifiers
| 745833-23-2 | |
| 404.33 | |
| FYSRKRZDBHOFAY-UHFFFAOYSA-N | |
| 10341154 | |
| NC(=O)N(C1=NC(=C(C=C1)C(N)=O)C1=C(F)C=C(F)C=C1)C1=C(F)C=CC=C1F |
| C19H12F4N4O2 | |
| MFCD11616590 | |
| 6-[(Aminocarbonyl)(2,6-difluorophenyl)amino]-2-(2,4-difluorophenyl)-3-pyridinecarboxamide, 2-(2,4-Difluorophenyl)-6-(1-[2,6-difluorophenyl)ureido]nicotinamide | |
| 2-(2,4-difluorophenyl)-6-[1-(2,6-difluorophenyl)carbamoylamino]pyridine-3-carboxamide |
Specifications
| 745833-23-2 | |
| C19H12F4N4O2 | |
| 25 mg | |
| FYSRKRZDBHOFAY-UHFFFAOYSA-N | |
| 2-(2,4-difluorophenyl)-6-[1-(2,6-difluorophenyl)carbamoylamino]pyridine-3-carboxamide | |
| 10341154 | |
| ≥98.0% (HPLC) | |
| VX-702 |
| White | |
| MFCD11616590 | |
| 6-[(Aminocarbonyl)(2,6-difluorophenyl)amino]-2-(2,4-difluorophenyl)-3-pyridinecarboxamide, 2-(2,4-Difluorophenyl)-6-(1-[2,6-difluorophenyl)ureido]nicotinamide | |
| NC(=O)N(C1=NC(=C(C=C1)C(N)=O)C1=C(F)C=C(F)C=C1)C1=C(F)C=CC=C1F | |
| 404.33 | |
| 404.32 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer (-20 °C)