Learn More
PolyCo™ VR™ Protective Gowns with Thumb Loops
Catalog No. p-4246865
Catalog No. 19125512 Supplier PolyCo™ Supplier No. 42500
Only null left
Gowns provide greater coverage for applications where the threat of biological or chemical contamination exist.
  • Glove-to-sleeve protection
  • Full-frontal coverage suitable for workplaces with biological or chemical hazards
  • Slips on over workers' heads and ties securely in the back for a quick and easy fit
  • Secures sleeves in place with thumb loop
  • Durable 4 mil thickness
Specifications
Length (English) 50 in.
Length (Metric) 127 cm
Product Type Controlled Environments Gown
For Use With (Application) Food Processing, Critical and Controlled Environments, Healthcare, Manufacturing
Closure Type Ties
Sterility Non-sterile
Seam Type Welded
Sleeve Style Full Sleeve with Thumb Loop
Certifications/Compliance OSHA 29, CFR Part 1910.1030