missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ VP-SFM AGT™ Medium

Catalog No. 12559027
Change view
Click to view available options
Quantity:
1 L
10 kg
10 L
100 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12559027 1 L
12559019 10 L
12559003 10 kg
12559001 100 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12559027 Supplier Gibco™ Supplier No. 12559027

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gibco VP-SFM AGT Medium is a flexible-format medium that is equivalent to VP-SFM with the addition of Advanced Granulation Technology for simplified preparation.

Gibco VP-SFM AGT Medium is a flexible-format medium that is equivalent to VP-SFM with the addition of Advanced Granulation Technology for simplified preparation. VP-SFM is a serum-free, animal-origin-free culture medium designed for growth of VERO cells and virus production. It is also suitable for the growth of COS-7, MDCK, BHK-21, and Hep2 cells and for recombinant protein production.

VP-SFM AGT™ is a serum-free, ultra-low protein medium in an easy-to-use granular format. It is designed for the growth of VERO cells for virus production and for the growth of COS-7, MDCK, BHK-21 and Hep-2 cells for recombinant protein production.
Advanced Granulation Technology = AGT

Specifications

Cell Line MDCK, VERO, HEp2, BHK-21, COS-7
Classification Animal Origin-free, Low Protein, Serum-free
Form Powder (AGT)
Product Type Serum Free Medium (SFM)
Serum Level Serum-free
With Additives Phenol Red, Sodium Pyruvate, Sodium Bicarbonate
Without Additives No Glutamine
Product Line Gibco
Quantity 1 L
Shipping Condition Wet Ice
Endotoxin Level Low
Format Securitainer Capped Vial
Included Antibiotics No antibiotics
Reconstituted Volume 1 L
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Culture Type Adherent Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.