missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ Vortexer Mixer, Blue

The Vortexer is a multi-purpose, and maximum convenience unit. This sleek and modern designed Vortexer Mixer is capable of meeting the mixing requirements of the most popular test tubes used in today’s lab without the need to swap out mixing head units.

Supplier:  Heathrow Scientific™ HS120209

 View more versions of this product

Catalog No. 14127520


Only null left

Description

Description

The innovative design makes maximum use of the space available, providing holding slots for no less than six different sizes of tube and a standard assay plate, while the center pad area can be used for quick hand-held mixing of a vast range of tubes and vessels.
  • Body made from durable ABS to resist most commonly used chemicals, and is easy to clean
  • Stable elastomeric device feet, provide vibration dampening and prevent movement during operation
  • Innovative head can be removed for cleaning purposes
  • US Design Patent No. D787,085, US Utility Patent No. 9895670
Specifications

Chemical Identifiers

3.7 mm
1000 to 3000 rpm
Vortexing unit
50/60 Hz
7.41 in.
6.7 x 7.5 in.
19.7 cm
5.51 in.
140 mm
4.5 in.
11.43 cm
5 kg
3.93 kg
1 x AC power cord, 1 x mixing head, 3 x elastic retention cords, 3 x central pads
3-Modes: 1000, 2000, and 3000 rpm
135 g
50 mL, 15 mL, 1.5/2 mL, 0.5 mL, and 0.2 mL
4.8 oz.
ABS Base, Nylon Platform
19 cm
7.68 in.
6.7 in.
17.1 cm
6.67 in.
17.1 cm
11.12 lb.
8.66 lb.
Blue
Brushless
5 Years

Specifications

3.7 mm
1000 to 3000 rpm
110/120 V
1 A
Vortexing unit
SGS-US-C, CE, CB Scheme, RoHS 2, PSE, WEEE
50/60 Hz
7.41 in.
6.7 x 7.5 in.
7.68 in.
6.7 in.
17.1 cm
6.67 in.
17.1 cm
11.12 lb.
8.66 lb.
Blue
Vortexer Mixer 110/120 V, US/NA Plug
Tubes, Assay Plates
Brushless
5 Years
135 g
110/120 V, 50/60 Hz
55 W
50 mL, 15 mL, 1.5/2 mL, 0.5 mL, and 0.2 mL
4.8 oz.
US Plug
ABS Base, Nylon Platform
19 cm
6.7 x 7.5 x 7.8 in.
19.7 cm
5.51 in.
140 mm
4.5 in.
11.43 cm
5 kg
3.93 kg
3 position slider (top) intermittent, (middle) off, (bottom) continuous
HS120209
1 x AC power cord, 1 x mixing head, 3 x elastic retention cords, 3 x central pads
3-Modes: 1000, 2000, and 3000 rpm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions