Learn More
BeLLCo Glass Manufactured from type 1 class a Borosilicate glass. Ideal for use to prepare solutions, and to contain. Flat bottom bulb with a elongated neck with permanent graduation line and marking area. Volumetric flasks are calibrated for a particular temperature
Supplier:  BeLLCo Glass 252500010
Encompass
Manufactured from type 1 class a Borosilicate glass. Ideal for use to prepare solutions, and to contain. Flat bottom bulb with a elongated neck with permanent graduation line and marking area. Volumetric flasks are calibrated for a particular temperature
Catalog No. 501945489
Only null left