missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Volcanic Tuff Specimen (Igneous Rock)

For use in exercises in igneous rock classification. EISCO Volcanic Tuff Specimen (Igneous Rock) is formed by ash and dust deposited during explosive volcanic eruptions that eventually become compacted and lithified, forming large packages of tuff. These

$112.00

Specifications

For Use With (Application) Exercises in igneous rock classification
Product Type Volcanic Tuff Study Specimen (Igneous Rock)
Width (English) Approx. 1 in.
Width (Metric) Approx. 3 cm
Packaging Clear labeled and barcoded bag
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
S24773
View Documents Promotion Details
Eisco™
ESNG0051PK12
12/Pk.
Pack of 12 Each for $112.00
Only null left
 
Description

Description

  • For use in classroom identification labs and exercises
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Distinct layers within tuff indicate separate eruption pulses or events
  • Comes in clear labeled and barcoded bag for storage
Specifications

Specifications

Exercises in igneous rock classification
Approx. 1 in.
Clear labeled and barcoded bag
Volcanic Tuff Study Specimen (Igneous Rock)
Approx. 3 cm
Videos
SDS
Documents

Documents