Learn More
ACROBiosystems Inc Recombinant Protein;Human VLDL R Protein;HEK293;100UG;VLR,VLDLR,RP11-320E16.1,CHRMQ1,FLJ35024,VLDLRCH,VLDL receptor
Catalog No. 501657773
Encompass
Recombinant Protein;Human VLDL R Protein;100UG;Human VLDL R, His Tag (VLR-H5227) is expressed from human 293 cells (HEK293). It contains AA Gly 28 - Ser 769 (Accession # P98155-2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 84.0 kDa. The protein migrates as 90-110 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.;Immobilized Human VLDL R, His Tag (Cat. No. VLR-H5227) at 5 g/mL (100 L/well) can bind Biotinylated Human PCSK9, His Tag (Cat. No. PC9-H82E7) with a linear range of 10-156 ng/mL (QC tested).
Catalog No. 501657773 Supplier ACROBiosystems Inc Supplier No. VLRH5227100UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;Human VLDL R Protein;100UG;Human VLDL R, His Tag (VLR-H5227) is expressed from human 293 cells (HEK293). It contains AA Gly 28 - Ser 769 (Accession # P98155-2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 84.0 kDa. The protein migrates as 90-110 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.;Immobilized Human VLDL R, His Tag (Cat. No. VLR-H5227) at 5 g/mL (100 L/well) can bind Biotinylated Human PCSK9, His Tag (Cat. No. PC9-H82E7) with a linear range of 10-156 ng/mL (QC tested).