missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaflow Tubing for Vivaflow Feed Reservoir

Catalog No. 10569128
Change view
Click to view available options
Diameter (English) Inner:
0.12 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English) Inner
10569128 0.12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10569128 Supplier Sartorius Supplier No. VFATD00011

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Vivaflow Tubing used with the Vivaflow feed reservoir, enables continuous diafiltration that is easy to set up, offers significant time savings, reduces reagent and water consumption, and avoids sample dilution.

  • Supports continuous diafiltration with Vivaflow cassettes
  • Securely connects to the Vivaflow feed reservoir
  • Works with Sartobind Lab for chromatography with a pump

Specifications

Color Code Transparent
Autoclavable Not Autoclavable
Diameter (English) Inner 0.12 in.
Diameter (Metric) Inner 3.1 mm
Material PVC
Diameter (English) Outer 0.24 in.
Diameter (Metric) Outer 6.3 mm
Overall Length (English) 39.4 in.
Overall Length (Metric) 1 m
Color Transparent
Product Type Tubing
Show More Show Less