missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitamin B12-Agarose, Saline Suspension, MilliporeSigma™ Supelco™
$907.05 - $907.05
Specifications
| MDL Number | MFCD00166171 |
|---|---|
| Synonym | Cyanocobalamin-Agarose |
| Physical Form | Saline Suspension |