missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies Pipette Tips: Low Retention
GREENER_CHOICE

Catalog No. 22146562
Change view
Click to view available options
Filtered:
Filtered
Non-filtered
Sterility:
Non-sterile
Sterile
Volume (Metric):
2 μL
25 μL
250 μL
1250 μL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filtered Sterility Volume (Metric)
22146562 Filtered Sterile 25 μL
22146563 Filtered Sterile 2 μL
22146560 Non-filtered Non-sterile 25 μL
22146568 Non-filtered Sterile 1250 μL
22146561 Non-filtered Sterile 25 μL
22146564 Non-filtered Non-sterile 250 μL
22146565 Non-filtered Sterile 250 μL
22146566 Filtered Sterile 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 22146562 Supplier VistaLab™ Technologies Supplier No. 40701332LR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with Ovation and MLA Brand pipetters

  • Sterile, Low Retention tips packaged in VistaRak: 5 racks of 192 tips each
  • Ideal for detergents, enzymes, viscous samples, foaming liquids, plasma, etc.
  • Use VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation pipettes
  • All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time

Specifications

Volume (Metric) 25 μL
Filtered Filtered
Sterility Sterile
Color Clear
Rack Color Blue
Autoclavable Not Autoclavable
Length (English) 1.7 in.
Thickness 5 in.
For Use With (Application) Ideal for detergents, enzymes, viscous samples, foaming liquids, plasma, TRIS, BSA and Tween, PCR or other biological applications
Sterilization Method e-beam
Format Rack
Tip Style Low Retention, Straight
Packaging VistaRack: 5 racks
Quantity 192 Tips/Rack
Product Type Pipette Tips
Color Code Blue
Disposable Single-use
For Use With (Equipment) Ovation
Graduated No
Material Polypropylene, Polyethylene (Filter)
Shape Cone
Show More Show Less