Learn More
Leica Microsystems Visoria P Digital Polarization Microscope for Industrial Applications (Transparent samples only)
This microscope uses polarized light to study the optical properties of materials and geological samples. The ergonomic design minimizes strain and improves comfort while working. Workflows can be streamlined with encoded functions, optimized light settings, and other features.
Supplier:  Leica Microsystems 111000013
Numéro de catalogue. 14-823-446
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Découvrez plus d'offres spéciales
Description
Description
  • Visoria P Digital Polarization Microscope with illumination management
  • 10 in. screen/tablet
  • Powered with Enersight microscope software (Desktop)
  • Flexacam i5 microscope camera, includes power supply
  • High-power white LED
  • Contrast methods (transmitted light): Quantitative polarization, Brightfield
  • Encoded nosepiece, 5x
  • Objectives (N PLAN POL): 2.5x, 5x, 10x, 20x, 40x
  • 360° rotatable polarization stage with verniers, 45° clickstop position and brake
  • Object guide without point counting
  • 360° Analyzer, Polarizer, Whole-wave plate f. compensator slot
  • 3-step focus drive with height-adjustable focus knobs
  • Dust protection cover
  • Quick start guide and user manual
  • Efficiency through encoded functions
  • More comfort with ergonomics
  • Visualize sample details with the right contrast
Spécifications
Spécifications
10X/22 mm
LED
Visoria P
Polarizing Microscope
Monitor
2.5X to 40X
Plan
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS