Learn More
Leica Microsystems Visoria M Digital Materials Microscope for Inspection and Measurement of Fine Details and Surface Structures
This microscope is suitable for metal, electronics, and polymer industries, and materials science labs. Ideal for inspecting and measuring fine details and surface structures of samples and workflows can be streamlined with encoded functions, optimized light settings, and other features.
Supplier:  Leica Microsystems 111000010
Catalog No. 14-823-443
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Visoria M Digital Materials Microscope with illumination management
  • 10 in. screen/tablet
  • Powered by Enersight microscope software platform (Desktop)
  • Flexacam i5 microscope camera, including power supply
  • High-power white LED
  • Incident light axis, 4-position
  • Contrast methods (incident light): Brightfield, Darkfield, Oblique illumination
  • Encoded nosepiece, 5x
  • Objectives (N PLAN EPI): 5x, 10x, 20x, 50x, 100x
  • Manual XY-stage (76 x 50 mm)
  • 3-step focus drive with height-adjustable focus knobs
  • Dust protection cover
  • Quick start guide and user manual
  • Efficiency through encoded functions
  • More comfort with ergonomics
  • Work without eyepieces by going digital
  • Visualize sample details with the right contrast
Specifications
Specifications
Monitor
5X to 100X
Plan
LED
Visoria M Digital
Digital Imaging Microscope
Product Suggestions
Product Suggestions