Learn More
Leica Microsystems Visoria B Laboratory Microscope for Hematopathology or Clinical Microbiology
Catalog No. 14823447
Change view
Click to view available options
Magnification Power:
5X to 40X
Catalog No. Magnification Power
14823447 5X to 40X
1 options
Catalog No. 14823447 Supplier Leica Microsystems Supplier No. 111000015
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This microscope is suitable for applications in life sciences and clinical laboratories to study hematopathology or clinical microbiology samples. Workflows can be streamlined with encoded functions, optimized light settings, and other features.
  • Visoria B Laboratory Microscope (Global IVD) with illumination management
  • Ergonomic tilting tube
  • Two eyepieces (10x), field of view 22 mm
  • High-power white LED
  • Contrast methods: Brightfield, Darkfield, Phase
  • Encoded nosepiece, 6x
  • Objectives (N PLAN): 5x, 10x, 20x, 40x, 50x, 100x
  • ErgoStage with Ceramic Plate, with specimen holder
  • 3-step focus drive with height-adjustable focus knobs
  • Condenser A 0.9 (P)
  • Immersion liquid, ISO 8036
  • Dust protection cover
  • Quick start guide and user manual
  • Efficiency through encoded functions
  • More comfort with ergonomics
  • Visualize sample details with the right contrast
Specifications
Eyepiece 10X/20 mm
Head Style Trinocular, Tilting
Lighting Type LED
Magnification Power 5X to 40X
Model Visoria B
Objective Types Plan
Product Type Binocular Microscope