Learn More
Leica Microsystems Visoria B Digital Laboratory Microscope for Histopathology or Cytopathology
This microscope is suitable for applications in life sciences and clinical laboratories to study histopathology or cytopathology samples. Workflows can be streamlined with encoded functions, optimized light settings, and other features.
Supplier:  Leica Microsystems 111000016
Catalog No. 14-823-448
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Visoria B Digital Laboratory Microscope (Global IVD) with illumination management
  • 10 in. screen/tablet
  • Powered by Enersight microscope software platform (Desktop)
  • Flexacam i5 microscope camera, includes power supply
  • High-power white LED
  • Contrast methods: Brightfield
  • Encoded nosepiece, 6x
  • Objectives (N PLAN): 2.5x, 5x, 10x, 20x, 40x
  • ErgoStage with ceramic plate, with specimen holder
  • 2-step focus drive with height-adjustable focus knobs
  • Condenser A 0.9 (P)
  • Dust protection cover
  • Quick start guide and user manual
  • Efficiency through encoded functions
  • More comfort with ergonomics
  • Work without eyepieces by going digital
  • Visualize sample details with the right contrast
Specifications
Specifications
Monitor
2.5X to 40X
Plan
LED
Visoria B
Biological Microscope
Product Suggestions
Product Suggestions