Learn More
MilliporeSigma™ Supelco™ Visiprep™ Large Volume Sampler
Catalog No. p-7190454
Change view
Click to view available options
For Use With (Equipment):
12, 20 or 60 mL SPE Tube
3 or 6 mL SPE Tube
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
111016309 3 or 6 mL SPE Tube
111016308 12, 20 or 60 mL SPE Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111016309 Supplier MilliporeSigma™ Supelco™ Supplier No. 57275
Only null left
For continuous 'hands off' direct transfer of multiple liquid samples. Large volume sampler enables you to transfer low viscosity samples directly from any sample container to conventional solid phase extraction tubes on a Visiprep SPE vacuum manifold.
Samplers consist of 1/8-inch PTFE tubing with a screw fitting at one end and a stainless steel weight at the other end.
To use the sampler, feed the weighted end into a sample container until it touches the bottom. Insert the tube adapters into the conditioned SPE tubes on your Visiprep manifold. Turn on the vacuum to the Visiprep manifold and open the manifold flow control valves, and the samples will be delivered to the SPE tubes. The flow rate of sample through each tube can be independently controlled by using the flow control valves.
Both the screw fittings and the PTFE tubing on the samplers are color coded for easy sample identification. Also fits Preppy SPE Vacuum Manifold.
Note: The Visiprep Large Volume Sampler can only be used with polypropylene SPE tubes. You must equip alternate manifold valves with long stem flow control knobs to accommodate 12 mL, 20 mL, or 60 mL SPE tubes.
Specifications
For Use With (Application) Food and Beverages
For Use With (Equipment) 3 or 6 mL SPE Tube
Product Line Visiprep
Type SPE Accessory
Includes 4 adapters
Visiprep is a trademark of Sigma-Aldrich Co. LLC