Learn More
Thermo Scientific™ VISION pro™ UV-Visible Spectrophotometer VISIONpro Software, Single license
Catalog No. 14386430
Change view
Click to view available options
Type:
VISIONpro Software, Single License
Catalog No. Type
14386430 VISIONpro Software, Single License
1 options
Catalog No. 14386430 Supplier Thermo Scientific™ Supplier No. 10040101
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Thermo Scientific™ VISION pro™ is full-featured instrument control software for applications from research to routine analysis.
Thermo Scientific™ VISION pro™ is full-featured instrument control software for applications from research to routine analysis. Powerful, versatile software offers advanced instrument control and data manipulation capabilities. Optimized for Thermo Scientific UV-Visible spectrophotometers, it brings a user-friendly, graphical environment to spectrophotometric analysis. Method setup, data collection, calculations, report presentation, regulatory compliance - all aspects of your analysis are easier with this software.
  • For use with Thermo Scientific Evolution™ 100, 160, 300 and 600 UV-Vis Spectrophotometers
  • Performs Scan, Fixed Wavelength Analysis, Quantification and Multi-Component Analysis
  • Produces spreadsheet results table
  • Arranges data easily with report composer
Includes:
User Manual and RS-232-C cable (Part No. 401317263061)
Requires:
System Requirements: Microsoft™ Windows™ 2000, XP, Vista
Specifications
Certifications/Compliance FDA guidelines, IEEE software engineering standards
Compatibility 10040101
Type VISIONpro Software, Single License
For Use With (Equipment) Evolution 300
Includes Fixed, Scan, Quant and MCA
System Requirements At least Windows XP