missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ViroCheck NanoParticle Reference Kit

Catalog No. V10425
Encompass
Change view
Click to view available options
Quantity:
25 tests kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V10425 25 tests kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V10425 Supplier Invitrogen™ Supplier No. V10425
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The ViroCheck NanoParticle Reference Kit is used for calibration of an intensity unit of fluorescence for flow cytometry analysis, and concentration analysis for virus and extracellular vesicles.

  • For calibration of an intensity unit of fluorescence for flow cytometry analysis and concentration analysis for virus and extracellular vesicles
  • Through the application of the NIST-assigned intensity standards for a given fluorophore, instrument sensitivity and accuracy can be determined
  • This permits data comparison between flow cytometry instruments
  • In combination with a biological standard, these particles can also be used to quantify the level of biomarker expression

ViroCheck NanoParticle Reference Kit features include:

  • NIST-assigned ERF values to 15 commonly used flow cytometry filter set channels
  • Enables normalization of the fluorescence intensity scale for instrument accuracy and sensitivity
  • NIST-verified nanoparticle concentration for counting of sub-micron biological samples such as viruses and extracellular vesicles (EV)
  • Facilitates instrument to instrument data comparison

Specifications

Quantity 25 tests kit
No. of Tests 25 tests
For Use With (Application) Flow Cytometry
Content And Storage
  • 100-nm FITC-like reference nanoparticles
  • 100-nm PE-like reference nanoparticles
  • 100-nm APC-like reference nanoparticles
  • 200-nm multicolor reference nanoparticles
  • 500-nm multicolor reference nanoparticles
For Use With (Equipment) Flow Cytometers
Regulatory Status RUO
Shipping Condition 2° to 8°C
Stability Do Not Freeze