missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Viral Production Medium

Catalog No. A4817903
Encompass
Change view
Click to view available options
Quantity:
1 L
6 L
10 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A4817903 10 L
A4817901 1 L
A4817902 6 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A4817903 Supplier Gibco™ Supplier No. A4817903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Viral Production Medium is a chemically defined, serum-free, and protein-free medium for growth and transfection of Viral Production Cells 2.0.

Gibco Viral Production Medium is a chemically defined, serum-free, and protein-free medium. It is animal origin-free and specially developed for growth and transfection of Viral Production Cells 2.0 (suspension-adapted HEK 293F-derived cells).

  • For adeno-associated virus (AAV) production as part of the AAV-MAX Helper-Free AAV Production System
  • Supports growth of suspension Viral Production Cells 2.0 to densities of over 1.2 x 107 cells/mL
  • Enables production of high AAV titers, greater than 5 x 1010 vg/mL (unpurified)
  • Supports growth and transfection of Viral Production Cells 2.0 from shake flask to bioreactor scales
  • Viral Production Medium requires supplementation with 4 mM GlutaMAX Supplement (Cat. No. 35050038) prior to use

Specifications

Cell Line Viral Production Cells 2.0
Cell Type HEK 293
Classification Serum-free, Animal Origin-free, Chemically-defined, Protein-free
Form Liquid
Product Type AAV Production Medium
Sterility Sterile-filtered
With Additives Glucose, Sodium Pyruvate, Sodium Pyruvate, GlutaMAX
Without Additives No GlutaMAX
For Use With (Application) AAV Production
Manufacturing Quality ISO 13485, MDSAP, FDA-registered
Product Line AAV-MAX
Quantity 10 L
Shipping Condition Room Temperature
Format Bag
pH 7
Regulatory Status RUO
Content And Storage 10 L bag; store at 2°C to 8°C, protected from 10 L BPC; Reagents should be stored at 2°C to 8°C, protected from light
Culture Type Suspension Cell Culture
Expression System Mammalian
Show More Show Less
Do I need to supplement the Viral Production Medium prior to use?

Yes, prior to using the Viral Production Medium, it should be supplemented with 4 mM GlutaMAX Supplement.

For Research Use Only. Not for use in diagnostic procedures.