missing translation for 'onlineSavingsMsg'
Learn More

Scigen VIP-Fixative Leak-Proof Specimen Containers Neutral Buffered 10% Formalin Filled

Safely transport, store and retrieve pathology specimens using Scigen’s Forma-Vial™ system. Containers from 7 mL to 7.5 Liters, filled with 10% neutral buffered formalin, OSHA compliant.

Supplier:  Scigen SGN2005

 View more versions of this product

Catalog No. 23730580

Only null left

Description

Description

  • Forma-Vial™ specimen containers are uniquely designed to be leak proof and protect against biohazardous or hazardous material release to the environment
  • Highly visible OSHA compliant labeling
  • Sizes range from 7mL dermatology containers to 7.5 Liters operating room organ buckets
  • Container openings range from 13mm to 300mm
  • Filled with 10% neutral buffered formalin to 50% of container’s capacity
  • Formalin waste designed to be treated in bulk using Neutralex or treated individually using Neutralexr tablets (patent pending).
  • Nonsterile containers marked with a lot code and expiration date, 4 year shelf life
  • Manufactured using ISO 13485 and FDA quality systems

Specifications

Chemical Identifiers

Yes
White
OSHA Compliant
Non-sterile
50% Filled Neutral Buffered 10% Formalin

Specifications

Specimen Container Vial
No
Non-sterile
960 mL, pre-filled 50%
Polypropylene
Lined Cap
Leakproof 960 mL specimen container
Histology/cytology specimen containers
100 mm
OSHA Compliant
5/Pk.
Yes
Polypropylene
32 oz., pre-filled 50%
White
120 mm
Translucent
114 mm
3.9 in.
50% Filled Neutral Buffered 10% Formalin
5/Cs.
Forma-Vial™ Specimen Containers
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions