missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ VIOS 160L 3 Door Inner Gas Tight Screen Door

Catalog No. 13642692
Change view
Click to view available options
Capacity (Metric):
160 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
13642692 160 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13642692 Supplier Thermo Scientific™ Supplier No. 51901144*

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

VIOS 160L 3 Door inner gas tight screen door

VIOS 160L 3 Door inner gas tight screen door

Specifications

Capacity (Metric) 160 L
For Use With (Equipment) Vios iDx 165
Type Inner Door
Quantity 3 Door