missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Violaxanthin, ≥95.0% (HPLC), MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 126-29-4 |
| Linear Formula | C40H56O4 |
| Synonym | (3 S,3 ′S,5 R,5 ′R,6 S,6 ′S)-5,6:5 ′,6 ′-Diepoxy-5,5 ′,6,6 ′-tetrahydro-beta,beta-carotene-3,3 ′-diol |
| Type | Analytical Standard |
| Quantity | 1 mg |
| Formula Weight | 600.87 |
| Percent Purity | ≥95.0% (HPLC) |
| Grade | Analytical Standard |