missing translation for 'onlineSavingsMsg'
Learn More

Vinyltrimethylsilane 97.0+%, TCI America™

Catalog No. p-7137912
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V006725ML 25 mL
V0067100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V006725ML Supplier TCI America Supplier No. V006725ML
Only null left

Chemical Identifiers

CAS 754-05-2
Molecular Formula C5H12Si
Molecular Weight (g/mol) 100.236
MDL Number MFCD00008606
InChI Key GCSJLQSCSDMKTP-UHFFFAOYSA-N
Synonym vinyltrimethylsilane, trimethyl vinyl silane, trimethylvinylsilane, silane, ethenyltrimethyl, trimethylsilyl ethylene, vinyl trimethylsilane, silane, trimethylvinyl, vinyltrimethysilane, trimethylsilyl ethene, ethenyl-trimethylsilane
PubChem CID 79102
IUPAC Name ethenyl(trimethyl)silane
SMILES C[Si](C)(C)C=C

Specifications

Color Yellow
Boiling Point 56°C
Quantity 25 mL
UN Number 1993
Formula Weight 100.24
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinyltrimethylsilane
EINECSNumber (2)-4007
TSCA Yes