missing translation for 'onlineSavingsMsg'
Learn More

Vinyltrimethylsilane, 97%

Catalog No. p-7043748
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0249809 10 g
AAL0249818 50 g
AAL0249830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0249809 Supplier Thermo Scientific Chemicals Supplier No. L0249809

CAS: 754-05-2 | C5H12Si | 100.236 g/mol

Vinyltrimethylsilane is used in semiconductor processing. It is involved in the Preparation of silyl-ethers by Rh(I) catalysis. Further, it is used in the preparation of trimethylsilylethyl sulfoxides which in the presence of fluoride ion generate sulfenate nucleophiles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vinyltrimethylsilane is used in semiconductor processing. It is involved in the Preparation of silyl-ethers by Rh(I) catalysis. Further, it is used in the preparation of trimethylsilylethyl sulfoxides which in the presence of fluoride ion generate sulfenate nucleophiles.

Solubility
Miscible with tetrahydrofuran, diethyl ether, benzene and dichloromethane.

Notes
Store in a cool place. Hygroscopic. Incompatible with strong oxidizing agents, strong acids and strong alkalis (bases).

Chemical Identifiers

CAS 754-05-2
Molecular Formula C5H12Si
Molecular Weight (g/mol) 100.236
MDL Number MFCD00008606
InChI Key GCSJLQSCSDMKTP-UHFFFAOYSA-N
Synonym vinyltrimethylsilane, trimethyl vinyl silane, trimethylvinylsilane, silane, ethenyltrimethyl, trimethylsilyl ethylene, vinyl trimethylsilane, silane, trimethylvinyl, vinyltrimethysilane, trimethylsilyl ethene, ethenyl-trimethylsilane
PubChem CID 79102
IUPAC Name ethenyl(trimethyl)silane
SMILES C[Si](C)(C)C=C

Specifications

Boiling Point 54°C to 56°C
Melting Point -132°C
Quantity 10 g
UN Number UN1993
Beilstein 956572
Flash Point −24°C (−11°F)
Solubility Information Miscible with tetrahydrofuran,diethyl ether,benzene and dichloromethane.
Formula Weight 100.24
Percent Purity 97%
Odor Mild
Refractive Index 1.391
Density 0.69
Chemical Name or Material Vinyltrimethylsilane
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 212-042-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only