missing translation for 'onlineSavingsMsg'
Learn More

Vinyltrimethoxysilane 98.0+%, TCI America™

Catalog No. p-7137897
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V004225ML 25 mL
V0042100ML 100 mL
V0042500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. V004225ML Supplier TCI America Supplier No. V004225ML
Only null left

Chemical Identifiers

CAS 2-7-2768
Molecular Formula C5H12O3Si
Molecular Weight (g/mol) 148.23
MDL Number MFCD00008605
InChI Key NKSJNEHGWDZZQF-UHFFFAOYSA-N
Synonym vinyltrimethoxysilane, trimethoxyvinylsilane, silane, ethenyltrimethoxy, trimethoxy vinyl silane, trimethoxysilyl ethene, vinyl trimethoxy silane, silane, trimethoxyvinyl, trimethoxysilyl ethylene, vtmo, vts-m
PubChem CID 76004
IUPAC Name ethenyl(trimethoxy)silane
SMILES CO[Si](C=C)(OC)OC

Specifications

Color Colorless
Boiling Point 123°C
Quantity 25 mL
UN Number 1993
Formula Weight 148.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinyltrimethoxysilane
EINECSNumber (2)-2066
RTECSNumber VV6770000
TSCA Yes