missing translation for 'onlineSavingsMsg'
Learn More

Vinyltrimethoxysilane, 98%

Catalog No. AAB2117622
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2117622 100 g
AAB2117636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2117622 Supplier Thermo Scientific Chemicals Supplier No. B2117622

CAS: 2768-02-7 | C5H12O3Si | 148.23 g/mol

Trimethoxyvinylsilane is used as a chemical intermediate in the production of other fine chemicals or as a monomer in the production of vinyl functional silicone polymers. The use of Trimethoxyvinylsilane in coatings is intended to improve bonding or adhesion between organic polymers and mineral surfaces, such as pigments, fillers, and glass or metal substrates. It is used in water-based decorative coating products. It is also used as a as cross-linking agent and adhesion promoter in commercially-available silicone sealant product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethoxyvinylsilane is used as a chemical intermediate in the production of other fine chemicals or as a monomer in the production of vinyl functional silicone polymers. The use of Trimethoxyvinylsilane in coatings is intended to improve bonding or adhesion between organic polymers and mineral surfaces, such as pigments, fillers, and glass or metal substrates. It is used in water-based decorative coating products. It is also used as a as cross-linking agent and adhesion promoter in commercially-available silicone sealant product.

Solubility
Solubility in water, 9.4 g/L at 20°C.

Notes
Moisture sensitive. Store away from oxidizing agents, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 2-7-2768
PubChem CID 76004

Specifications

Boiling Point 122°C to 124°C
Melting Point <-40°C
Quantity 100 g
Molecular Formula C5H12O3Si
Linear Formula CH2=CHSi(OCH3)3
MDL Number MFCD00008605
UN Number UN1993
Beilstein 1099136
Flash Point 23°C (73°F)
Synonym vinyltrimethoxysilane, trimethoxyvinylsilane, silane, ethenyltrimethoxy, trimethoxy vinyl silane, trimethoxysilyl ethene, vinyl trimethoxy silane, silane, trimethoxyvinyl, trimethoxysilyl ethylene, vtmo, vts-m
InChI Key NKSJNEHGWDZZQF-UHFFFAOYSA-N
SMILES CO[Si](C=C)(OC)OC
IUPAC Name ethenyl(trimethoxy)silane
Molecular Weight (g/mol) 148.23
Formula Weight 148.24
Percent Purity 98%
Odor Fruit-like
Refractive Index 1.392
Sensitivity Moisture sensitive
Density 0.97
Chemical Name or Material Vinyltrimethoxysilane
Show More Show Less
Hazard Category H226-H332-H335-H373
Hazard Statement GHS H Statement
H318-H226-H332-H335-H315
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P271-P280-P303+P361+P353-P304+P340-P312-P314-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 220-449-8
RTECSNumber VV6770000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only