missing translation for 'onlineSavingsMsg'
Learn More

Vinyltrimethoxysilane, 98%, AcroSeal™

Catalog No. AC433061000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC433061000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC433061000 Supplier Thermo Scientific Chemicals Supplier No. 433061000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2768-02-7 | C5H12O3Si | 148.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2-7-2768
Molecular Formula C5H12O3Si
Molecular Weight (g/mol) 148.24
MDL Number MFCD00008605
InChI Key NKSJNEHGWDZZQF-UHFFFAOYSA-N
Synonym vinyltrimethoxysilane, trimethoxyvinylsilane, silane, ethenyltrimethoxy, trimethoxy vinyl silane, trimethoxysilyl ethene, vinyl trimethoxy silane, silane, trimethoxyvinyl, trimethoxysilyl ethylene, vtmo, vts-m
PubChem CID 76004
IUPAC Name ethenyl(trimethoxy)silane
SMILES CO[Si](C=C)(OC)OC

Specifications

Color Colorless
Density 0.9680g/mL
Boiling Point 123.0°C
Flash Point 23°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula H2C=CHSi(OCH3)3
Refractive Index 1.3915 to 1.3935
Quantity 100 mL
Specific Gravity 0.968
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in most common organic solvents
Viscosity 0.6 mm2/s (25°C)
Formula Weight 148.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Vinyltrimethoxysilane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest
EINECSNumber 220-449-8

RUO – Research Use Only