missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vinylphosphonic Acid 95.0+%, TCI America™
$236.80 - $1466.98
Chemical Identifiers
| CAS | 1746-03-8 |
|---|---|
| Molecular Formula | C2H3O3P |
| Molecular Weight (g/mol) | 106.02 |
| MDL Number | MFCD00043866 |
| InChI Key | ZTWTYVWXUKTLCP-UHFFFAOYSA-L |
| Synonym | vinylphosphonic acid, phosphonic acid, ethenyl-, homopolymer, vinyl-phosphonicaci, ethenephosphonicacid, ethenylphosphonicacid, ethenyl-phosphonicaci, ethylenephosphonicacid, phosphonic acid, p-ethenyl-, homopolymer, acmc-1bvr6, poly vinylphosphonic acid |
| PubChem CID | 168725 |
| IUPAC Name | ethenylphosphonate |
| SMILES | [O-]P([O-])(=O)C=C |
Chemical Identifiers
| 1746-03-8 | |
| 106.02 | |
| ZTWTYVWXUKTLCP-UHFFFAOYSA-L | |
| 168725 | |
| [O-]P([O-])(=O)C=C |
| C2H3O3P | |
| MFCD00043866 | |
| vinylphosphonic acid, phosphonic acid, ethenyl-, homopolymer, vinyl-phosphonicaci, ethenephosphonicacid, ethenylphosphonicacid, ethenyl-phosphonicaci, ethylenephosphonicacid, phosphonic acid, p-ethenyl-, homopolymer, acmc-1bvr6, poly vinylphosphonic acid | |
| ethenylphosphonate |
Specifications
| 1746-03-8 | |
| 98°C | |
| MFCD00043866 | |
| 3265 | |
| ZTWTYVWXUKTLCP-UHFFFAOYSA-L | |
| ethenylphosphonate | |
| 168725 | |
| ≥95.0% (T) | |
| Vinylphosphonic Acid |
| Yellow | |
| C2H3O3P | |
| 25 g | |
| vinylphosphonic acid, phosphonic acid, ethenyl-, homopolymer, vinyl-phosphonicaci, ethenephosphonicacid, ethenylphosphonicacid, ethenyl-phosphonicaci, ethylenephosphonicacid, phosphonic acid, p-ethenyl-, homopolymer, acmc-1bvr6, poly vinylphosphonic acid | |
| [O-]P([O-])(=O)C=C | |
| 106.02 | |
| 108.03 | |
| Liquid |
Safety and Handling
RTECSNumber : SZ7903500
TSCA : Yes