Learn More
Vinylmagnesium bromide, 1M solution in 2-MeTHF, AcroSeal™
CAS: 1826-67-1 | C2H3BrMg | 131.26 g/mol
Supplier: Thermo Scientific Chemicals 445978000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Vinylmagnesium bromide | |
| 0.9650g/mL | |
| AcroSeal™ Glass bottle | |
| CH2CHMgBr | |
| 800 mL | |
| vinylmagnesium bromide, bromo ethenyl magnesium, vinyl magnesium bromide, magnesium, bromoethenyl, vinylmagnesium bromide solution, vinylmagnesium bromide solution, 1.0 m in thf, bromovinylmagnesium, bromo vinyl magnesium, vinyl magnesiumbromide | |
| XHHHAXOHMKAOSL-UHFFFAOYSA-M | |
| magnesium;ethene;bromide | |
| 74584 | |
| 1M Solution in 2-MeTHF |
| 1826-67-1,96-47-9 | |
| −11°C | |
| C2H3BrMg | |
| MFCD00000042 | |
| 0.965 | |
| Solubility in water: reacts | |
| Br[Mg]C=C | |
| 131.26 | |
| 131.25 |
Chemical Identifiers
| 1826-67-1 | |
| 131.26 | |
| XHHHAXOHMKAOSL-UHFFFAOYSA-M | |
| 74584 | |
| Br[Mg]C=C |
| C2H3BrMg | |
| MFCD00000042 | |
| vinylmagnesium bromide, bromo ethenyl magnesium, vinyl magnesium bromide, magnesium, bromoethenyl, vinylmagnesium bromide solution, vinylmagnesium bromide solution, 1.0 m in thf, bromovinylmagnesium, bromo vinyl magnesium, vinyl magnesiumbromide | |
| magnesium;ethene;bromide |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Reacts violently with water.<b
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture
GHS Signal Word: Danger
EINECSNumber : 217-375-3
RUO – Research Use Only