missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vinylbenzyl trimethylammonium chloride, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 26616-35-3 |
|---|---|
| Molecular Formula | C12H18ClN |
| Molecular Weight (g/mol) | 211.73 |
| MDL Number | MFCD00080690 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC422560250
|
Thermo Scientific Chemicals
422560250 |
25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC422561000
|
Thermo Scientific Chemicals
422561000 |
100g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Phase transfer catalystChemical Identifiers
| 26616-35-3 | |
| C12H18ClN | |
| 211.73 | |
| MFCD00080690 |
Specifications
| 26616-35-3 | |
| 96% min. (Argentometry) | |
| C12H18ClN | |
| MFCD00080690 | |
| 211.73 | |
| 97% |
| Authentic | |
| Glass bottle | |
| CH2CHC6H4CH2N(CH3)3Cl | |
| 25g | |
| 211.73 | |
| Vinylbenzyl trimethylammonium chloride |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS Signal Word: Warning
EINECSNumber : 247-850-
RUO â Research Use Only