missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Vinylacetic acid, tech., 90%, unstabilized

CAS: 625-38-7 | C4H6O2 | 86.09 g/mol

$178.24 - $508.22

Chemical Identifiers

CAS 625-38-7
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00002782
InChI Key PVEOYINWKBTPIZ-UHFFFAOYSA-N
Synonym 3-butenoic acid, vinylacetic acid, vinyl acetic acid, acetic acid, ethenyl, beta-butenoic acid, .beta.-butenoic acid, 2-propenylcarboxylic acid, allylic acid, vinylessigsaeure, 3-butenoicacid
PubChem CID 32743
ChEBI CHEBI:35897
IUPAC Name but-3-enoic acid
SMILES OC(=O)CC=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC158830250
SDS View Documents
Thermo Scientific Chemicals
158830250
25 g Glass bottle
Each for $178.24
Only null left
 
AC158831000
SDS View Documents
Thermo Scientific Chemicals
158831000
100 g Glass bottle
Each for $508.22
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

625-38-7
86.09
PVEOYINWKBTPIZ-UHFFFAOYSA-N
32743
but-3-enoic acid
C4H6O2
MFCD00002782
3-butenoic acid, vinylacetic acid, vinyl acetic acid, acetic acid, ethenyl, beta-butenoic acid, .beta.-butenoic acid, 2-propenylcarboxylic acid, allylic acid, vinylessigsaeure, 3-butenoicacid
CHEBI:35897
OC(=O)CC=C

Specifications

625-38-7
Colorless to Yellow
163°C
Authentic
C4H6O2
H2C=CHCH2CO2H
25 g
1.01
Solubility in water: soluble. Other solubilities: soluble in most common organic solvents
OC(=O)CC=C
86.09
CHEBI:35897
90%
Liquid
-39°C
1.0100g/mL
65°C
Glass bottle
1.4239 to 1.4259
MFCD00002782
02, 407
3-butenoic acid, vinylacetic acid, vinyl acetic acid, acetic acid, ethenyl, beta-butenoic acid, .beta.-butenoic acid, 2-propenylcarboxylic acid, allylic acid, vinylessigsaeure, 3-butenoicacid
PVEOYINWKBTPIZ-UHFFFAOYSA-N
but-3-enoic acid
32743
86.09
Technical
Vinylacetic acid, tech., 90%, unstabilized
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-892-5

SDS
Documents

Documents

RUO – Research Use Only