missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Vinylacetic acid, tech., 90%, unstabilized

Catalog No. AC158831000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158831000 100 g Glass bottle
AC158830250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC158831000 Supplier Thermo Scientific Chemicals Supplier No. 158831000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 625-38-7 | C4H6O2 | 86.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 625-38-7
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00002782
InChI Key PVEOYINWKBTPIZ-UHFFFAOYSA-N
Synonym 3-butenoic acid, vinylacetic acid, vinyl acetic acid, acetic acid, ethenyl, beta-butenoic acid, .beta.-butenoic acid, 2-propenylcarboxylic acid, allylic acid, vinylessigsaeure, 3-butenoicacid
PubChem CID 32743
ChEBI CHEBI:35897
IUPAC Name but-3-enoic acid
SMILES OC(=O)CC=C

Specifications

Melting Point -39°C
Color Colorless to Yellow
Density 1.0100g/mL
Boiling Point 163°C
Flash Point 65°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4239 to 1.4259
Linear Formula H2C=CHCH2CO2H
Quantity 100 g
Beilstein 02, 407
Specific Gravity 1.01
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most common organic solvents
Formula Weight 86.09
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Vinylacetic acid, tech., 90%, unstabilized
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-892-5

RUO – Research Use Only