missing translation for 'onlineSavingsMsg'
Learn More

Vinylacetic acid, 96%

Catalog No. AAL0755722
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0755722 100 g
AAL0755714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0755722 Supplier Thermo Scientific Chemicals Supplier No. L0755722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 625-38-7 | C4H6O2 | 86.09 g/mol

Vinylacetic acid used in the preparation of strained nitroso Diels-Aldrer adducts which undergo a domino ROC metathesis in the presence of Ru-carbene catalysts and olefins. It is used in the synthesis of polyethylene glycolated boltorn coatings. It was used to study the inhibition of the enzyme-catalyzed conversion of D-tyrosyl- phenylalanyl-glycine to D-tyrosyl-phenylaline amide by non-peptide carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vinylacetic acid used in the preparation of strained nitroso Diels-Aldrer adducts which undergo a domino ROC metathesis in the presence of Ru-carbene catalysts and olefins. It is used in the synthesis of polyethylene glycolated boltorn coatings. It was used to study the inhibition of the enzyme-catalyzed conversion of D-tyrosyl- phenylalanyl-glycine to D-tyrosyl-phenylaline amide by non-peptide carboxylic acids.

Solubility
Fully miscible.

Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents. Incompatible with oxidizing agents, heat, Bases, Reducing agents.

Chemical Identifiers

CAS 625-38-7
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00002782
InChI Key PVEOYINWKBTPIZ-UHFFFAOYSA-N
Synonym 3-butenoic acid, vinylacetic acid, vinyl acetic acid, acetic acid, ethenyl, beta-butenoic acid, .beta.-butenoic acid, 2-propenylcarboxylic acid, allylic acid, vinylessigsaeure, 3-butenoicacid
PubChem CID 32743
ChEBI CHEBI:35897
IUPAC Name but-3-enoic acid
SMILES OC(=O)CC=C

Specifications

Melting Point -39°C
Density 1.012
Boiling Point 162°C to 163°C
Flash Point 65°C (149°F)
Refractive Index 1.424
Linear Formula H2C=CHCH2CO2H
Quantity 100 g
UN Number UN3265
Beilstein 1699159
Solubility Information Fully miscible.
Formula Weight 86.09
Percent Purity 96%
Chemical Name or Material Vinylacetic acid
Show More Show Less
Hazard Category H227-H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 210-892-5
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only