missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Vinyl Tubing and Connector Kits

Resist fruit acids, lye, alcohol, greases, and oils.

Supplier:  Fisherbrand™ 8108514EMD

 View more versions of this product

Catalog No. S37110


Only null left
Explore available promotions

Description

Description

  • Clear, flexible vinyl tubing
  • Nontoxic and tasteless
  • Can be used at temperatures from −33° to +100°C (−27° to +212°F) at low pressures
  • Tensile strength: 2000psi (13.8MPa)
  • Durometer hardness: Shore A, 65

Specifications

Specifications

U.S. Pharmacopeia XIX, Class VI, and National Formulary Vol. 14 StandardFDA criteria: 178.3740, 178.2010, and 175.300 (Tubing only)
9.5 mm
3/8 in.
50 ft.
0.125 in.
1/16 in.
Vinyl
15.9 mm
15.2 m
3.2mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions